ENA413166008 Enamine

(2-methanesulfonyl-2-methylpropyl)hydrazine hydrochloride

  • MDL number MFCD27500728
  • Cas Number : 1803588-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142584
Availability details for
Quantity
  • Availability
  • Add to Cart