ENA413166011 Enamine

ethyl 4-chloro-8-methoxy-3-methylquinoline-2-carboxylate

  • MDL number MFCD26936283
  • Cas Number : 1803592-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142614
Availability details for
Quantity
  • Availability
  • Add to Cart