ENA413166017 Enamine

2-ethylbenzamide

  • MDL number MFCD18824517
  • Cas Number : 67832-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140487
Availability details for
Quantity
  • Availability
  • Add to Cart