ENA413166024 Enamine

3-methyl-1-(propan-2-yl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD06805443
  • Cas Number : 1006453-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40...42
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139874

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139874
Availability details for
Quantity
  • Availability
  • Add to Cart