ENA413166027 Enamine

ethyl 5-acetyl-2,6-dimethylpyridine-3-carboxylate

  • MDL number MFCD27500699
  • Cas Number : 30428-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140503
Availability details for
Quantity
  • Availability
  • Add to Cart