ENA413166031 Enamine

1,2,3-tribromo-5-nitrobenzene

  • MDL number MFCD02657489
  • Cas Number : 3460-20-6
  • Molecular Weight : 359.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2,3-tribromo-5-nitrobenzene
Inchi Code 1S/C6H2Br3NO2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H
Inchi Key AKLYERMMCABILW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-139609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139609
Availability details for
Quantity
  • Availability
  • Add to Cart