ENA413166043 Enamine

4,6-dimethyl-2-(methylsulfanyl)pyridine-3-carbonitrile

  • MDL number MFCD00124685
  • Cas Number : 72456-86-1
  • Molecular Weight : 178.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-140548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140548
Availability details for
Quantity
  • Availability
  • Add to Cart