ENA413166046 Enamine

2-cyclopentyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD16698580
  • Cas Number : 1247889-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140556
Availability details for
Quantity
  • Availability
  • Add to Cart