ENA413166049 Enamine

3-hydroxy-2-[2-(naphthalen-1-yl)acetamido]butanoic acid

  • MDL number MFCD13271459

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140559
Availability details for
Quantity
  • Availability
  • Add to Cart