ENA413166052 Enamine

methyl 2-amino-2-(2,4-dimethoxyphenyl)acetate hydrochloride

  • MDL number MFCD26936107
  • Cas Number : 1607016-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140565
Availability details for
Quantity
  • Availability
  • Add to Cart