ENA413166053 Enamine

1-[1-(azetidin-3-yl)-1H-1,2,3-triazol-4-yl]ethan-1-ol dihydrochloride

  • MDL number MFCD26936056
  • Cas Number : 1798032-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139897
Availability details for
Quantity
  • Availability
  • Add to Cart