ENA413166055 Enamine

methyl 2-[1-(aminomethyl)cyclohexyl]acetate hydrochloride

  • MDL number MFCD26936110
  • Cas Number : 63562-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140577
Availability details for
Quantity
  • Availability
  • Add to Cart