ENA413166056 Enamine

4-(cyclobutylmethyl)morpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD26936111
  • Cas Number : 1796895-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140578
Availability details for
Quantity
  • Availability
  • Add to Cart