ENA413166057 Enamine

benzyl(3,3-dimethylbutan-2-yl)amine

  • MDL number MFCD20547703
  • Cas Number : 356518-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140580
Availability details for
Quantity
  • Availability
  • Add to Cart