ENA413166059 Enamine

tert-butyl N-[(1R)-1-cyanopropyl]carbamate

  • MDL number MFCD24603303
  • Cas Number : 1604264-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139902
Availability details for
Quantity
  • Availability
  • Add to Cart