ENA413166063 Enamine

5-(2-nitrophenyl)-2,3-dihydro-1,3,4-oxadiazol-2-one

  • MDL number MFCD19673273
  • Cas Number : 83725-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140602

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140602
Availability details for
Quantity
  • Availability
  • Add to Cart