ENA413166067 Enamine

2-methyl-4-[(2S)-pyrrolidin-2-yl]-1,3-thiazole

  • MDL number MFCD18645719
  • Cas Number : 1272743-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139914
Availability details for
Quantity
  • Availability
  • Add to Cart