ENA413166072 Enamine

2-(2-methoxyphenyl)-2-(propan-2-yloxy)ethan-1-amine

  • MDL number MFCD16063030
  • Cas Number : 1247681-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140629
Availability details for
Quantity
  • Availability
  • Add to Cart