ENA413166075 Enamine

benzyl N-(chlorosulfonyl)carbamate

  • MDL number MFCD19200062
  • Cas Number : 89979-13-5
  • Molecular Weight : 249.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-140636

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-140636
pictograms GHS05
hstatements H314,H318
Availability details for
Quantity
  • Availability
  • Add to Cart