ENA413166077 Enamine

4-ethyl 2-pyridin-2-ylmethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD08526939
  • Cas Number : 923767-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140638

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-140638
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart