ENA413166079 Enamine

methyl 3-amino-3-{[1,1'-biphenyl]-4-yl}propanoate

  • MDL number MFCD12487142
  • Cas Number : 479591-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140640
Availability details for
Quantity
  • Availability
  • Add to Cart