ENA413166082 Enamine

3-(2H-1,3-benzodioxol-4-yl)propanoic acid

  • MDL number MFCD20639318
  • Cas Number : 20974-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140643

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140643
Availability details for
Quantity
  • Availability
  • Add to Cart