ENA413166083 Enamine

1-(2-hydroxyethyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD21922803
  • Cas Number : 1520462-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140645
Availability details for
Quantity
  • Availability
  • Add to Cart