ENA413166086 Enamine

3-(3-chlorophenyl)imidazolidine-2,4-dione

  • MDL number MFCD04213335
  • Cas Number : 42351-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140648
Availability details for
Quantity
  • Availability
  • Add to Cart