ENA413166087 Enamine

1-(1-bromo-2-methylpropyl)-2,4-difluorobenzene

  • MDL number MFCD12789984
  • Cas Number : 1249632-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140650
Availability details for
Quantity
  • Availability
  • Add to Cart