ENA413166101 Enamine

3-[(3,5-difluorophenyl)carbamoyl]-2,2-dimethylpropanoic acid

  • MDL number MFCD20990149
  • Cas Number : 1485705-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140670
Availability details for
Quantity
  • Availability
  • Add to Cart