ENA413166102 Enamine

1-(2,5-dimethylphenyl)-2-[(propan-2-yl)amino]ethan-1-ol

  • MDL number MFCD17218723
  • Cas Number : 1267612-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140671
Availability details for
Quantity
  • Availability
  • Add to Cart