ENA413166103 Enamine

2-iodo-N-(2,2,2-trifluoroethyl)acetamide

  • MDL number MFCD20414803
  • Cas Number : 1343302-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140672
Availability details for
Quantity
  • Availability
  • Add to Cart