ENA413166107 Enamine

2-(tert-butoxy)ethanimidamide hydrochloride

  • MDL number MFCD26936121
  • Cas Number : 14401-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140695

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-140695
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart