ENA413166110 Enamine

2-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)ethan-1-ol

  • MDL number MFCD26936059
  • Cas Number : 121941-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139981
Availability details for
Quantity
  • Availability
  • Add to Cart