ENA413166111 Enamine

1,3-diethyl 2-[(4-fluorophenyl)methyl]-2-methylpropanedioate

  • MDL number MFCD26936124
  • Cas Number : 1108187-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140700
Availability details for
Quantity
  • Availability
  • Add to Cart