ENA413166112 Enamine

5-(4-chloro-2-fluorophenyl)-3-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD10693315
  • Cas Number : 1152498-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140705
Availability details for
Quantity
  • Availability
  • Add to Cart