ENA413166114 Enamine

N-[4-(hydroxymethyl)phenyl]pent-4-enamide

  • MDL number MFCD20414090
  • Cas Number : 1344062-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140715
Availability details for
Quantity
  • Availability
  • Add to Cart