ENA413166115 Enamine

5-{[(2,6-dichloropyridin-4-yl)amino]methylidene}-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD26395999
  • Cas Number : 863785-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140717

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140717
Availability details for
Quantity
  • Availability
  • Add to Cart