ENA413166122 Enamine

2-amino-6-chloro-4-fluorophenol

  • MDL number MFCD16742728
  • Cas Number : 260253-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140735

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140735
Availability details for
Quantity
  • Availability
  • Add to Cart