ENA413166129 Enamine

1-[(4-methylphenyl)sulfanyl]propan-2-amine

  • MDL number MFCD20392350
  • Cas Number : 104864-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140748
Availability details for
Quantity
  • Availability
  • Add to Cart