ENA413166130 Enamine

5-amino-1-(3-methylbutyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD14661902
  • Cas Number : 159979-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139614
Availability details for
Quantity
  • Availability
  • Add to Cart