ENA413166131 Enamine

ethyl 2-(ethylamino)-3-phenylpropanoate hydrochloride

  • MDL number MFCD26936129
  • Cas Number : 92196-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140750
Availability details for
Quantity
  • Availability
  • Add to Cart