ENA413166137 Enamine

2-{2-[(2-methylphenyl)methyl]-1H-1,3-benzodiazol-1-yl}acetic acid

  • MDL number MFCD13665563
  • Cas Number : 1097800-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248...250
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140756
Availability details for
Quantity
  • Availability
  • Add to Cart