ENA413166142 Enamine

2-amino-2-(thiophen-3-yl)ethan-1-ol

  • MDL number MFCD12827184
  • Cas Number : 1183538-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40...42
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140761

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140761
Availability details for
Quantity
  • Availability
  • Add to Cart