ENA413166146 Enamine

2-[(1-hydroxybutan-2-yl)amino]propanoic acid

  • MDL number MFCD16088582
  • Cas Number : 1501996-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140768
Availability details for
Quantity
  • Availability
  • Add to Cart