ENA413166149 Enamine

2-chloro-6-(4-methylpiperidin-1-yl)pyrazine

  • MDL number MFCD10614662
  • Cas Number : 885704-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140771
Availability details for
Quantity
  • Availability
  • Add to Cart