ENA413166150 Enamine

2-[(6-chloropyrazin-2-yl)amino]ethan-1-ol

  • MDL number MFCD13562417
  • Cas Number : 1147998-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140772

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140772
Availability details for
Quantity
  • Availability
  • Add to Cart