ENA413166151 Enamine

bis(N-ethyl-N-methylguanidine); sulfuric acid

  • MDL number MFCD26936132
  • Cas Number : 1798040-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2H2SO4
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140776
Availability details for
Quantity
  • Availability
  • Add to Cart