ENA413166154 Enamine

3-(benzyloxy)-2,2-dimethylcyclobutan-1-amine

  • MDL number MFCD21757638
  • Cas Number : 1518855-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140779
Availability details for
Quantity
  • Availability
  • Add to Cart