ENA413166159 Enamine

6-amino-5-hydroxypyridine-2-carboxylic acid

  • MDL number MFCD11109881
  • Cas Number : 61548-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140784
Availability details for
Quantity
  • Availability
  • Add to Cart