ENA413166169 Enamine

2-(thiophene-2-carbonyl)-1H-imidazole

  • MDL number MFCD02679401
  • Cas Number : 610279-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140812
Availability details for
Quantity
  • Availability
  • Add to Cart