ENA413166176 Enamine

3-methyl-2-sulfanylbenzoic acid

  • MDL number MFCD20638540
  • Cas Number : 77149-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140820
Availability details for
Quantity
  • Availability
  • Add to Cart