ENA413166182 Enamine

methyl 4-bromo-3-(chlorosulfonyl)-5-fluorobenzoate

  • MDL number MFCD16084851
  • Cas Number : 1249508-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140838
Availability details for
Quantity
  • Availability
  • Add to Cart