ENA413166190 Enamine

methyl 2-(4-chlorophenyl)-2-oxoacetate

  • MDL number MFCD00157150
  • Cas Number : 37542-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140848
Availability details for
Quantity
  • Availability
  • Add to Cart